Product Details

SNP ID
rs145435553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:109335223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTCTTTTTTAAAATTTAAG[C/T]CAACTCTTATTCAACTTTTCTTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC285638 PubMed Links

Gene Details

Gene
LOC285638
Gene Name
uncharacterized LOC285638
There are no transcripts associated with this gene.

Gene
PJA2
Gene Name
praja ring finger ubiquitin ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014819.4 4431 UTR 3 NP_055634.3
XM_017010099.1 4431 UTR 3 XP_016865588.1

View Full Product Details