Product Details

SNP ID
rs145483785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90399520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATGCCTTGAGTATTTCTTCAT[A/G]GGGATCTCTTTCCAATATCCAGTCT
Phenotype
MIM: 602907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CETN3 PubMed Links

Gene Details

Gene
CETN3
Gene Name
centrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297765.1 423 Missense Mutation CAT,TAT H100Y NP_001284694.1
NM_001297768.1 423 Missense Mutation CAT,TAT H100Y NP_001284697.1
NM_004365.3 423 Missense Mutation CAT,TAT H100Y NP_004356.2

View Full Product Details