Product Details

SNP ID
rs145507265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177512488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAACCCATGCCCTTGGGCCCCAG[A/G]CTCTTACCATAGTTCTCAATCTCCT
Phenotype
MIM: 608170 MIM: 611435 MIM: 615813
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX41 PubMed Links

Gene Details

Gene
DDX41
Gene Name
DEAD-box helicase 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321732.1 Intron NP_001308661.1
NM_001321830.1 Intron NP_001308759.1
NM_016222.3 Intron NP_057306.2
Gene
DOK3
Gene Name
docking protein 3
There are no transcripts associated with this gene.

Gene
FAM193B
Gene Name
family with sequence similarity 193 member B
There are no transcripts associated with this gene.

View Full Product Details