Product Details

SNP ID
rs145673143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180844484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGCCATGGTTAAGATGGTAAAA[C/G]TCTGGCTCTCACCAACCCTTCCTAC
Phenotype
MIM: 610281
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00847 PubMed Links

Gene Details

Gene
LINC00847
Gene Name
long intergenic non-protein coding RNA 847
There are no transcripts associated with this gene.

Gene
ZFP62
Gene Name
ZFP62 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172638.1 4151 Intron NP_001166109.1
NM_152283.4 4151 Intron NP_689496.4
XM_017009710.1 4151 Intron XP_016865199.1
XM_017009711.1 4151 Intron XP_016865200.1
XM_017009712.1 4151 Intron XP_016865201.1
XM_017009713.1 4151 Intron XP_016865202.1
XM_017009714.1 4151 Intron XP_016865203.1
XM_017009715.1 4151 Intron XP_016865204.1
XM_017009716.1 4151 UTR 3 XP_016865205.1
XM_017009717.1 4151 Intron XP_016865206.1
XM_017009718.1 4151 Intron XP_016865207.1
XM_017009719.1 4151 Intron XP_016865208.1
XM_017009720.1 4151 Intron XP_016865209.1
XM_017009721.1 4151 Intron XP_016865210.1

View Full Product Details