Product Details
- SNP ID
-
rs146207929
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:134765908 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTAATTCAAGTACTTAAGTGAAA[A/G]TGGTTATTTAACATTTGGTCTTTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDX46
PubMed Links
Gene Details
- Gene
- DDX46
- Gene Name
- DEAD-box helicase 46
View Full Product Details