Product Details

SNP ID
rs146231810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:24488026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCAGGGTGCCGATATCAAAGGC[C/T]TGGGTGTCCTCCTCTCCACCACCCT
Phenotype
MIM: 604555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH10 PubMed Links

Gene Details

Gene
CDH10
Gene Name
cadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317222.1 2007 Silent Mutation NP_001304151.1
NM_001317224.1 2007 Silent Mutation NP_001304153.1
NM_006727.4 2007 Silent Mutation NP_006718.2
XM_011513923.2 2007 Missense Mutation XP_011512225.1
XM_017008915.1 2007 Missense Mutation XP_016864404.1

View Full Product Details