Product Details

SNP ID
rs146496148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157181064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCCTCGAACTTTAAAGTCCG[A/C]TTCTTTGTGTTAACCAAAGCCAGCC
Phenotype
MIM: 186973
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITK PubMed Links

Gene Details

Gene
ITK
Gene Name
IL2 inducible T-cell kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005546.3 169 Silent Mutation CGA,CGC R29R NP_005537.3
XM_017009443.1 169 Intron XP_016864932.1

View Full Product Details