Product Details

SNP ID
rs146547482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148451236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATTGTGTATGGGCAGTTTCTC[A/G]AGTTCCTGATGATGTCCCCTGTGCA
Phenotype
MIM: 608533 MIM: 602164
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO38 PubMed Links

Gene Details

Gene
FBXO38
Gene Name
F-box protein 38
There are no transcripts associated with this gene.

Gene
HTR4
Gene Name
5-hydroxytryptamine receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000870.6 1236 Intron NP_000861.1
NM_001040169.2 1236 Silent Mutation NP_001035259.1
NM_001040172.2 1236 Intron NP_001035262.2
NM_001040173.2 1236 Intron NP_001035263.1
NM_001286410.1 1236 Intron NP_001273339.1
NM_199453.3 1236 UTR 3 NP_955525.1

View Full Product Details