Product Details

SNP ID
rs146664105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:205709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTTCTTGCATCACCTGGGCC[C/T]GTTCCTGTTCCAGAAGCTTCTTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC127 PubMed Links

Gene Details

Gene
CCDC127
Gene Name
coiled-coil domain containing 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145265.2 471 Missense Mutation CAG,CGG Q124R NP_660308.1

View Full Product Details