Product Details

SNP ID
rs147182349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163439386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAATTGAGTCTGCACACGATAAT[A/G]GCCTCAGAATGACTGCAAGACTAAG
Phenotype
MIM: 601578
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNG1 PubMed Links

Gene Details

Gene
CCNG1
Gene Name
cyclin G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004060.3 364 Missense Mutation AGC,GGC S44G NP_004051.1
NM_199246.1 364 Missense Mutation AGC,GGC S44G NP_954854.1

View Full Product Details