Product Details

SNP ID
rs147476583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150054183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGCTCACTGCTGCTGCTGCC[A/G]CTGCCACCGCTTCTGCTGCTGCTCG
Phenotype
MIM: 164770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSF1R PubMed Links

Gene Details

Gene
CSF1R
Gene Name
colony stimulating factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288705.1 3097 Silent Mutation AGC,AGT S935S NP_001275634.1
NM_005211.3 3097 Silent Mutation AGC,AGT S935S NP_005202.2
Gene
HMGXB3
Gene Name
HMG-box containing 3
There are no transcripts associated with this gene.

View Full Product Details