Product Details
- SNP ID
-
rs147510643
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:50428224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGAGAGGTGGACTTGTAAAAGGC[A/G]AATCTATAAGAGAAAGAACACATGA
- Phenotype
-
MIM: 615669
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EMB
PubMed Links
Gene Details
- Gene
- EMB
- Gene Name
- embigin
View Full Product Details