Product Details

SNP ID
rs147836081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34915749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGCGAGGCAAGATGGCGGCAA[C/G]CAAGAGGAAACGGCGTGGAGGCTTT
Phenotype
MIM: 603153
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRIX1 PubMed Links

Gene Details

Gene
BRIX1
Gene Name
BRX1, biogenesis of ribosomes
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018321.3 35 Missense Mutation ACC,AGC T4S NP_060791.3
Gene
RAD1
Gene Name
RAD1 checkpoint DNA exonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002853.3 35 Intron NP_002844.1

View Full Product Details