Product Details
- SNP ID
-
rs148398791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:19473344 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTGTGTCGTTGCTGAATCCAGC[A/G]AGCTGATAGACCCAGCTTCTGATCT
- Phenotype
-
MIM: 603019
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH18
PubMed Links
Gene Details
- Gene
- CDH18
- Gene Name
- cadherin 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001167667.1 |
3246 |
UTR 3 |
|
|
NP_001161139.1 |
NM_001291956.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
NP_001278885.1 |
NM_001291957.1 |
3246 |
UTR 3 |
|
|
NP_001278886.1 |
NM_004934.3 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
NP_004925.1 |
XM_005248228.3 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_005248285.1 |
XM_006714435.3 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_006714498.1 |
XM_011513928.2 |
3246 |
Missense Mutation |
TCG,TTG |
S686L |
XP_011512230.1 |
XM_011513929.2 |
3246 |
Missense Mutation |
TCG,TTG |
S584L |
XP_011512231.1 |
XM_011513930.2 |
3246 |
UTR 3 |
|
|
XP_011512232.1 |
XM_017008924.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864413.1 |
XM_017008925.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864414.1 |
XM_017008926.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864415.1 |
XM_017008927.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864416.1 |
XM_017008928.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864417.1 |
XM_017008929.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864418.1 |
XM_017008930.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864419.1 |
XM_017008931.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864420.1 |
XM_017008932.1 |
3246 |
Missense Mutation |
TCG,TTG |
S752L |
XP_016864421.1 |
XM_017008933.1 |
3246 |
Missense Mutation |
TCG,TTG |
S529L |
XP_016864422.1 |
XM_017008934.1 |
3246 |
Missense Mutation |
TCG,TTG |
S529L |
XP_016864423.1 |
XM_017008935.1 |
3246 |
UTR 3 |
|
|
XP_016864424.1 |
XM_017008936.1 |
3246 |
UTR 3 |
|
|
XP_016864425.1 |
XM_017008937.1 |
3246 |
UTR 3 |
|
|
XP_016864426.1 |
XM_017008938.1 |
3246 |
UTR 3 |
|
|
XP_016864427.1 |
XM_017008939.1 |
3246 |
UTR 3 |
|
|
XP_016864428.1 |
XM_017008940.1 |
3246 |
UTR 3 |
|
|
XP_016864429.1 |
XM_017008941.1 |
3246 |
UTR 3 |
|
|
XP_016864430.1 |
View Full Product Details