Product Details

SNP ID
hCV171693831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140801281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCGCAGCCTCGGAGGTGGGGAG[C/G]GGCCAGCTCCACTACTCCGTCTCTG
Phenotype
MIM: 606307 MIM: 606308 MIM: 606309 MIM: 606310
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
There are no transcripts associated with this gene.

Gene
PCDHA2
Gene Name
protocadherin alpha 2
There are no transcripts associated with this gene.

Gene
PCDHA3
Gene Name
protocadherin alpha 3
There are no transcripts associated with this gene.

Gene
PCDHA4
Gene Name
protocadherin alpha 4
There are no transcripts associated with this gene.

View Full Product Details