Product Details

SNP ID
rs150258538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128259560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTTCTTAAGCTCCTTCTTCTT[G/T]TAGAGAGGGATGCTAGTGATTTCCA
Phenotype
MIM: 612570
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBN2 PubMed Links

Gene Details

Gene
FBN2
Gene Name
fibrillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001999.3 9236 Nonsense Mutation TAA,TAC *2878Y NP_001990.2
XM_017009228.1 9236 Nonsense Mutation TAA,TAC *2827Y XP_016864717.1

View Full Product Details