Product Details

SNP ID
rs150789725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73554956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCAACGACAGTGCACTTTCTC[A/G]ACCTTTTCCTAAAAGTTGGGGATCA
Phenotype
MIM: 605787
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 1160 Nonsense Mutation CGA,TGA R215* NP_075526.1
XM_005248560.2 1160 Nonsense Mutation CGA,TGA R147* XP_005248617.1
XM_017009678.1 1160 Nonsense Mutation CGA,TGA R54* XP_016865167.1

View Full Product Details