Product Details

SNP ID
rs150819002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90458514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACATACTAATATTTCCAAAAAT[C/T]ATTTAAGTTACAAGGGTCACTGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBLAC2 PubMed Links

Gene Details

Gene
MBLAC2
Gene Name
metallo-beta-lactamase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203406.1 3969 UTR 3 NP_981951.1

View Full Product Details