Product Details

SNP ID
hCV171852237
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31270081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGGGATGGTGGGCTGGGAAGA[C/T]GGCTCTGGGAAAGGAGGAGAAGGTG
Phenotype
MIM: 142840
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HLA-C PubMed Links
Additional Information
For this assay, SNP(s) [rs9264621] are located under a probe and SNP(s) [rs35424484] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HLA-C
Gene Name
major histocompatibility complex, class I, C
There are no transcripts associated with this gene.

View Full Product Details