Product Details

SNP ID
rs77625743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29673368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACCCAGATGGACGCGGCGGTGA[T/C]GACTTAGTCCAGAAGCATCACAGTA
Phenotype
MIM: 159465 MIM: 612192
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 709 Intron NP_001008229.1
NM_001008229.2 709 Intron NP_001008230.1
NM_001170418.1 709 Intron NP_001163889.1
NM_002433.4 709 Intron NP_002424.3
NM_206809.3 709 Intron NP_996532.2
NM_206810.3 709 Intron NP_996533.2
NM_206811.3 709 Intron NP_996534.2
NM_206812.3 709 Intron NP_996535.2
NM_206814.5 709 Intron NP_996537.3
XM_005249131.3 709 Intron XP_005249188.1
Gene
ZFP57
Gene Name
ZFP57 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109809.2 709 Missense Mutation CAT,CGT H248R NP_001103279.2
XM_006715087.3 709 Missense Mutation CAT,CGT H176R XP_006715150.1
XM_011514570.2 709 Missense Mutation CAT,CGT H248R XP_011512872.1

View Full Product Details