Product Details

SNP ID
rs137897838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33319043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGTGAATGGGGGGTTTGGGAC[A/T]GCCGGGCTGGAGAAGGGATGCAGAG
Phenotype
MIM: 603186 MIM: 601962 MIM: 611439
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DAXX PubMed Links

Gene Details

Gene
DAXX
Gene Name
death domain associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141969.1 1171 Missense Mutation CAG,CTG Q706L NP_001135441.1
NM_001141970.1 1171 Missense Mutation CAG,CTG Q718L NP_001135442.1
NM_001254717.1 1171 Missense Mutation CAG,CTG Q631L NP_001241646.1
NM_001350.4 1171 Missense Mutation CAG,CTG Q706L NP_001341.1
XM_005248860.3 1171 Missense Mutation CAG,CTG Q338L XP_005248917.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
ZBTB22
Gene Name
zinc finger and BTB domain containing 22
There are no transcripts associated with this gene.

View Full Product Details