Product Details
- SNP ID
-
rs138200747
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35500115 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCAGTTCTATGAGGCTCTCCAGC[A/G]TCTTGTTCTGCCAGCGCACCAGCAG
- Phenotype
-
MIM: 603170
MIM: 602280
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TEAD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41270076] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TEAD3
- Gene Name
- TEA domain transcription factor 3
There are no transcripts associated with this gene.
- Gene
- TULP1
- Gene Name
- tubby like protein 1
View Full Product Details