Product Details

SNP ID
rs138216669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36971506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACCCACCCACAAGGCCCCCCGA[A/G]AAGGGGCAGAACTGCAGGAATGTAC
Phenotype
MIM: 610449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTCH1 PubMed Links

Gene Details

Gene
MTCH1
Gene Name
mitochondrial carrier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271641.1 Intron NP_001258570.1
NM_014341.2 Intron NP_055156.1
XM_005248978.1 Intron XP_005249035.1
XM_005248979.1 Intron XP_005249036.1
XM_011514450.1 Intron XP_011512752.1
XM_017010642.1 Intron XP_016866131.1
XM_017010643.1 Intron XP_016866132.1
Gene
PI16
Gene Name
peptidase inhibitor 16
There are no transcripts associated with this gene.

View Full Product Details