Product Details

SNP ID
rs138283476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43484559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAATAGGAAAAGGCTTTGTTCT[A/G]TTAAATCTAAAATAAGCCTTTTGAT
Phenotype
MIM: 612658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TJAP1 PubMed Links

Gene Details

Gene
TJAP1
Gene Name
tight junction associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146016.1 Intron NP_001139488.1
NM_001146017.1 Intron NP_001139489.1
NM_001146018.1 Intron NP_001139490.1
NM_001146019.1 Intron NP_001139491.1
NM_001146020.1 Intron NP_001139492.1
NM_080604.2 Intron NP_542171.2
XM_006715249.1 Intron XP_006715312.1
XM_006715250.3 Intron XP_006715313.1
XM_006715251.2 Intron XP_006715314.1
XM_006715252.2 Intron XP_006715315.1
XM_006715254.3 Intron XP_006715317.1
XM_006715256.1 Intron XP_006715319.1
XM_006715257.2 Intron XP_006715320.1
XM_006715259.3 Intron XP_006715322.1
XM_006715260.1 Intron XP_006715323.1
XM_006715261.1 Intron XP_006715324.1
XM_006715262.2 Intron XP_006715325.1
XM_006715263.2 Intron XP_006715326.1
XM_006715265.1 Intron XP_006715328.1
XM_006715266.1 Intron XP_006715329.1
XM_006715267.2 Intron XP_006715330.1
XM_006715268.2 Intron XP_006715331.1
XM_006715269.2 Intron XP_006715332.1
XM_006715270.1 Intron XP_006715333.1
XM_011514995.1 Intron XP_011513297.1
XM_011514996.1 Intron XP_011513298.1
XM_017011488.1 Intron XP_016866977.1
XM_017011489.1 Intron XP_016866978.1
XM_017011490.1 Intron XP_016866979.1
XM_017011491.1 Intron XP_016866980.1
XM_017011492.1 Intron XP_016866981.1
XM_017011493.1 Intron XP_016866982.1

View Full Product Details