Product Details

SNP ID
rs138607789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:97142432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGCTATGTTGCTAGCTTCCCAA[A/T]CCAACTGTTTCTAACCAAATAAAGC
Phenotype
MIM: 615614
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL32 PubMed Links

Gene Details

Gene
KLHL32
Gene Name
kelch like family member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286250.1 4847 Intron NP_001273179.1
NM_001286251.1 4847 Intron NP_001273180.1
NM_001286252.1 4847 Intron NP_001273181.1
NM_001286254.2 4847 Intron NP_001273183.1
NM_001323252.1 4847 Intron NP_001310181.1
NM_001323253.1 4847 Intron NP_001310182.1
NM_001323254.1 4847 Intron NP_001310183.1
NM_001323255.1 4847 Intron NP_001310184.1
NM_001323256.1 4847 Intron NP_001310185.1
NM_001323257.1 4847 Intron NP_001310186.1
NM_001323258.1 4847 Intron NP_001310187.1
NM_001323259.1 4847 Intron NP_001310188.1
NM_001323260.1 4847 Intron NP_001310189.1
NM_001323261.1 4847 Intron NP_001310190.1
NM_001323262.1 4847 Intron NP_001310191.1
NM_001323263.1 4847 Intron NP_001310192.1
NM_001323264.1 4847 Intron NP_001310193.1
NM_052904.3 4847 Intron NP_443136.2
XM_005266813.4 4847 UTR 3 XP_005266870.1
XM_017010228.1 4847 UTR 3 XP_016865717.1
XM_017010229.1 4847 UTR 3 XP_016865718.1
Gene
MMS22L
Gene Name
MMS22 like, DNA repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198468.2 4847 UTR 3 NP_940870.2
XM_006715430.1 4847 UTR 3 XP_006715493.1
XM_006715432.3 4847 Intron XP_006715495.1
XM_011535670.2 4847 Intron XP_011533972.1
XM_011535671.2 4847 Intron XP_011533973.1
XM_011535672.2 4847 Intron XP_011533974.1
XM_011535673.2 4847 Intron XP_011533975.1
XM_011535674.2 4847 Intron XP_011533976.1
XM_011535675.2 4847 Intron XP_011533977.1
XM_011535676.2 4847 Intron XP_011533978.1
XM_011535677.2 4847 Intron XP_011533979.1
XM_011535678.2 4847 Intron XP_011533980.1
XM_011535679.2 4847 Intron XP_011533981.1
XM_011535680.2 4847 Intron XP_011533982.1
XM_017010656.1 4847 Intron XP_016866145.1
XM_017010657.1 4847 Intron XP_016866146.1

View Full Product Details