Product Details

SNP ID
rs138799104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159035644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACATTTTTACATATTGCACAAAT[C/G]CAGGTACACAGAGACTATCTGATGC
Phenotype
MIM: 609667
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAGAP PubMed Links

Gene Details

Gene
TAGAP
Gene Name
T-cell activation RhoGTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278733.1 2711 UTR 3 NP_001265662.1
NM_054114.4 2711 UTR 3 NP_473455.2
NM_138810.3 2711 Intron NP_620165.1
NM_152133.2 2711 UTR 3 NP_687034.1

View Full Product Details