Product Details

SNP ID
rs36211074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29397257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTAAGCCATTGCTAAAGCTGGC[C/T]TGTGGGAACACTGAGCTTAATCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR12D2 PubMed Links

Gene Details

Gene
OR12D2
Gene Name
olfactory receptor family 12 subfamily D member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013936.3 619 Silent Mutation GCC,GCT A186A NP_039224.2
Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 619 Intron NP_110503.3
XM_011514929.2 619 Intron XP_011513231.1
XM_011514930.2 619 Intron XP_011513232.1

View Full Product Details