Product Details

SNP ID
rs138951019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26123893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCGGGGCGGGAGCAGACTTGGC[C/T]GGCTCAGGCATCTTAAAACACCAGA
Phenotype
MIM: 602794 MIM: 602847
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AC PubMed Links

Gene Details

Gene
HIST1H2AC
Gene Name
histone cluster 1, H2ac
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003512.3 Intron NP_003503.1
Gene
HIST1H2BC
Gene Name
histone cluster 1, H2bc
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003526.2 Intron NP_003517.2

View Full Product Details