Product Details
- SNP ID
-
rs139039407
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:27147182 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCTCGCGCCAAGGCCAAGACCCG[C/T]TCTTCTCGGGCTGGGCTTCAGTTCC
- Phenotype
-
MIM: 615013
MIM: 615045
MIM: 602833
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST1H2AH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141253641,rs6919033] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST1H2AH
- Gene Name
- histone cluster 1, H2ah
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080596.2 |
|
Intron |
|
|
NP_542163.1 |
- Gene
- HIST1H2BK
- Gene Name
- histone cluster 1, H2bk
- Gene
- HIST1H4I
- Gene Name
- histone cluster 1, H4i
There are no transcripts associated with this gene.
- Gene
- MIR3143
- Gene Name
- microRNA 3143
There are no transcripts associated with this gene.
View Full Product Details