Product Details

SNP ID
rs139464075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10896170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAACAAAACAAGCCCTACAGAA[A/G]GGAACATCTTTAGTCCTGTACCGTA
Phenotype
MIM: 616799
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYCP2L PubMed Links

Gene Details

Gene
SYCP2L
Gene Name
synaptonemal complex protein 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040274.2 Intron NP_001035364.2

View Full Product Details