Product Details

SNP ID
rs139552047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:166929594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCCAAAACTTGGGCATCAATG[C/T]TTGGTCTTTTTAGGTGGGGGATAGA
Phenotype
MIM: 612944
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASET2 PubMed Links

Gene Details

Gene
RNASET2
Gene Name
ribonuclease T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003730.4 1172 Silent Mutation AAA,AAG K255K NP_003721.2
XM_017011397.1 1172 Intron XP_016866886.1
XM_017011398.1 1172 Intron XP_016866887.1
XM_017011399.1 1172 UTR 3 XP_016866888.1

View Full Product Details