Product Details

SNP ID
rs139823805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42185796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCTTTCTTCAGCTGGTAAATT[C/T]CCTGCCAAAGAAAACTCAGCTGCAT
Phenotype
MIM: 600364 MIM: 602275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1A PubMed Links

Gene Details

Gene
GUCA1A
Gene Name
guanylate cyclase activator 1A
There are no transcripts associated with this gene.

Gene
GUCA1B
Gene Name
guanylate cyclase activator 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002098.5 495 Missense Mutation GAA,GGA E120G NP_002089.4

View Full Product Details