Product Details

SNP ID
rs139989472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158188386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTAAAGAGTCTGAGTAGAAAAA[C/T]AATGGTTTTCAAGAAACTGAAACTT
Phenotype
MIM: 608780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2H5 PubMed Links

Gene Details

Gene
GTF2H5
Gene Name
general transcription factor IIH subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207118.2 Intron NP_997001.1
XM_017010862.1 Intron XP_016866351.1

View Full Product Details