Product Details

SNP ID
rs140688078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43523556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACAGGTATGTGGCTGCACGGGG[C/G]TGGGAGGGCTTGTGGGAGAAGGGCT
Phenotype
MIM: 610060 MIM: 607845 MIM: 609775
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR1C PubMed Links

Gene Details

Gene
POLR1C
Gene Name
RNA polymerase I subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318876.1 4138 Intron NP_001305805.1
NM_203290.3 4138 Intron NP_976035.1
XM_011515000.2 4138 Intron XP_011513302.1
Gene
XPO5
Gene Name
exportin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020750.2 4138 UTR 3 NP_065801.1
Gene
YIPF3
Gene Name
Yip1 domain family member 3
There are no transcripts associated with this gene.

View Full Product Details