Product Details

SNP ID
rs141408930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30915784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTACAGGGGAGACCTTTTCCAT[A/G]TGTATCCCACCTCCCAATGTCACTG
Phenotype
MIM: 601760 MIM: 612802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2H4 PubMed Links

Gene Details

Gene
GTF2H4
Gene Name
general transcription factor IIH subunit 4
There are no transcripts associated with this gene.

Gene
VARS2
Gene Name
valyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167733.2 587 Missense Mutation ATA,ATG I1M NP_001161205.1
NM_001167734.1 587 Missense Mutation ATA,ATG I171M NP_001161206.1
NM_020442.5 587 Missense Mutation ATA,ATG I141M NP_065175.4

View Full Product Details