Product Details

SNP ID
rs141584461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28431078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGGTAACTAGTTATGTTAACAA[C/T]GGGTTTGTCTTAATTAGGGGCGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN23 PubMed Links

Gene Details

Gene
ZSCAN23
Gene Name
zinc finger and SCAN domain containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012455.1 2336 Intron NP_001012458.1
XM_005248950.2 2336 UTR 3 XP_005249007.1
XM_011514406.2 2336 Intron XP_011512708.1
XM_011514408.2 2336 Intron XP_011512710.1
XM_011514410.2 2336 Intron XP_011512712.1
XM_017010479.1 2336 Intron XP_016865968.1

View Full Product Details