Product Details

SNP ID
rs141778979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49960301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTATGCCTTTCTTTTCAAAACAT[A/G]TCATCTTCTTCATACAATTTCTCAG
Phenotype
MIM: 615243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEFB113 PubMed Links

Gene Details

Gene
DEFB113
Gene Name
defensin beta 113
There are no transcripts associated with this gene.

Gene
DEFB114
Gene Name
defensin beta 114
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037499.1 201 Silent Mutation GAC,GAT D67D NP_001032588.1

View Full Product Details