Product Details

SNP ID
rs141879172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:130028615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACAACAGACCTTAATTCCAAT[A/G]TATGTGTGATTCAGAGTACTTATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
L3MBTL3 PubMed Links

Gene Details

Gene
L3MBTL3
Gene Name
l(3)mbt-like 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007102.2 Intron NP_001007103.1
NM_032438.2 Intron NP_115814.1
XM_005267161.4 Intron XP_005267218.1
XM_006715576.3 Intron XP_006715639.1
XM_006715578.3 Intron XP_006715641.1
XM_011536179.2 Intron XP_011534481.1
XM_011536180.2 Intron XP_011534482.1
XM_011536181.2 Intron XP_011534483.1
XM_011536183.2 Intron XP_011534485.1
XM_011536184.2 Intron XP_011534486.1
XM_017011355.1 Intron XP_016866844.1
XM_017011356.1 Intron XP_016866845.1

View Full Product Details