Product Details

SNP ID
rs142002674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44276846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACAGATCTTCTGTTAGATGACA[A/G]CCGTGCCGTGGGCCCCGCGTTGGGC
Phenotype
MIM: 186975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCTE1 PubMed Links

Gene Details

Gene
TCTE1
Gene Name
t-complex-associated-testis-expressed 1
There are no transcripts associated with this gene.

Gene
TMEM151B
Gene Name
transmembrane protein 151B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137560.1 2020 UTR 3 NP_001131032.1

View Full Product Details