Product Details
- SNP ID
-
rs142541451
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:52832887 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCAGAGGGAAGCTGGAGATAAGA[A/C]TCGAGTCAAGCTCTTCCACGTAGTA
- Phenotype
-
MIM: 607605
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GSTA5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs55815585] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GSTA5
- Gene Name
- glutathione S-transferase alpha 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153699.1 |
588 |
Missense Mutation |
|
|
NP_714543.1 |
View Full Product Details