Product Details

SNP ID
rs142600886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73223753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCTTCCCAGGGGAGATCAGT[C/T]TCCGGTGTACGGTGGCAGGCTAACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KHDC1L PubMed Links

Gene Details

Gene
KHDC1L
Gene Name
KH domain containing 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126063.2 Intron NP_001119535.1

View Full Product Details