Product Details

SNP ID
rs142908633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41798824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATCATAGGCAAAGGTCTCTTT[C/G]TCAAGAGAGGAGAGCATGTCCCTGC
Phenotype
MIM: 616168
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TOMM6 PubMed Links

Gene Details

Gene
TOMM6
Gene Name
translocase of outer mitochondrial membrane 6
There are no transcripts associated with this gene.

Gene
USP49
Gene Name
ubiquitin specific peptidase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286554.1 1878 Missense Mutation GAC,GAG D592E NP_001273483.1
NM_018561.4 1878 Missense Mutation GAC,GAG D592E NP_061031.2

View Full Product Details