Product Details

SNP ID
rs143629937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109392859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATCTCCACTGTCAGTAATTCT[A/G]CACATATGTATTGGTCTTTCATTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIL6 PubMed Links

Gene Details

Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111298.2 926 Silent Mutation TGC,TGT C327C NP_001104768.2
NM_001286360.1 926 Silent Mutation TGC,TGT C269C NP_001273289.1
NM_001286361.1 926 UTR 3 NP_001273290.1
NM_173672.4 926 Silent Mutation TGC,TGT C301C NP_775943.1
XM_011535765.2 926 Silent Mutation TGC,TGT C295C XP_011534067.1
XM_011535766.2 926 Intron XP_011534068.1
XM_011535767.2 926 Intron XP_011534069.1
XM_011535769.2 926 Intron XP_011534071.1
XM_017010774.1 926 UTR 3 XP_016866263.1

View Full Product Details