Product Details

SNP ID
rs143752545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110392940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATCTCGCGCAAGGAGCTCTGT[C/T]TGCAGTCGCACGCCTGGCCTGTAGG
Phenotype
MIM: 124450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDO PubMed Links

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003649.2 772 Silent Mutation CAA,CAG Q315Q NP_003640.2
NM_004032.2 772 Silent Mutation CAA,CAG Q256Q NP_004023.2

View Full Product Details