Product Details

SNP ID
rs143961414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:157400723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCTTCTGCTCAAAACACTCCT[A/G]CCCTATTCCAGCCACGAACCTATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC14 PubMed Links

Gene Details

Gene
ZDHHC14
Gene Name
zinc finger DHHC-type containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024630.2 Intron NP_078906.2
NM_153746.1 Intron NP_714968.1
XM_005267146.3 Intron XP_005267203.1
XM_011536120.2 Intron XP_011534422.1
XM_011536123.2 Intron XP_011534425.1
XM_017011307.1 Intron XP_016866796.1
XM_017011308.1 Intron XP_016866797.1
XM_017011309.1 Intron XP_016866798.1
XM_017011310.1 Intron XP_016866799.1
XM_017011311.1 Intron XP_016866800.1
XM_017011312.1 Intron XP_016866801.1
XM_017011313.1 Intron XP_016866802.1

View Full Product Details