Product Details

SNP ID
rs144573705
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36954845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCACAGGCCCCGTTGGAGCCCTC[A/G]CAGATGAGGAGAAACGTTTGATGGT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PI16 PubMed Links

Gene Details

Gene
PI16
Gene Name
peptidase inhibitor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199159.1 308 Missense Mutation ACA,GCA T29A NP_001186088.1
NM_153370.2 308 Missense Mutation ACA,GCA T29A NP_699201.2
XM_005248917.2 308 Missense Mutation ACA,GCA T29A XP_005248974.1
XM_011514375.2 308 Missense Mutation ACA,GCA T29A XP_011512677.1
XM_017010430.1 308 Missense Mutation ACA,GCA T29A XP_016865919.1

View Full Product Details