Product Details

SNP ID
rs144632729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGTCGAGCAGCACCCGTTCGGC[C/G]AGGCCGCCTACCCCGCCGACGGCGC
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 784 Missense Mutation CAG,GAG Q146E NP_001025034.1
NM_001317342.1 784 Missense Mutation CAG,GAG Q165E NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 784 Intron NP_001189479.1
NM_182552.4 784 Intron NP_872358.4
XM_011535682.2 784 Intron XP_011533984.1
XM_011535684.2 784 Intron XP_011533986.1
XM_011535685.2 784 Intron XP_011533987.1
XM_011535687.2 784 Intron XP_011533989.1
XM_011535688.2 784 Intron XP_011533990.1
XM_011535689.2 784 Intron XP_011533991.1
XM_011535690.2 784 Intron XP_011533992.1
XM_011535691.2 784 Intron XP_011533993.1
XM_011535692.2 784 Intron XP_011533994.1
XM_011535693.2 784 Intron XP_011533995.1
XM_011535694.2 784 Intron XP_011533996.1
XM_011535696.2 784 Intron XP_011533998.1
XM_011535697.2 784 Intron XP_011533999.1
XM_017010658.1 784 Intron XP_016866147.1
XM_017010659.1 784 Intron XP_016866148.1
XM_017010660.1 784 Intron XP_016866149.1
XM_017010661.1 784 Intron XP_016866150.1
XM_017010662.1 784 Intron XP_016866151.1
XM_017010663.1 784 Intron XP_016866152.1
XM_017010664.1 784 Intron XP_016866153.1
XM_017010665.1 784 Intron XP_016866154.1
XM_017010666.1 784 Intron XP_016866155.1
XM_017010667.1 784 Intron XP_016866156.1
XM_017010668.1 784 Intron XP_016866157.1
XM_017010669.1 784 Intron XP_016866158.1
XM_017010670.1 784 Intron XP_016866159.1
XM_017010671.1 784 Intron XP_016866160.1
XM_017010672.1 784 Intron XP_016866161.1
XM_017010673.1 784 Intron XP_016866162.1
XM_017010674.1 784 Intron XP_016866163.1
XM_017010675.1 784 Intron XP_016866164.1

View Full Product Details