Product Details
- SNP ID
-
rs144782925
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29397519 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCATGGACCAGGACCGGATTGTT[A/G]CCATCATGTACACTGTGGTCACTCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR12D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs36211075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR12D2
- Gene Name
- olfactory receptor family 12 subfamily D member 2 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013936.3 |
881 |
Missense Mutation |
ACC,GCC |
T274A |
NP_039224.2 |
- Gene
- OR5V1
- Gene Name
- olfactory receptor family 5 subfamily V member 1
View Full Product Details