Product Details

SNP ID
rs144838471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169703648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTGTCATACCTATGAAAATTC[G/T]TCAGTTATTAAATAACTTTGTCAGT
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 1537 UTR 3 NP_001025034.1
NM_001317342.1 1537 UTR 3 NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018288.3 1537 Intron NP_060758.2
NM_133325.2 1537 Intron NP_579866.2
XM_017010998.1 1537 Intron XP_016866487.1
Gene
WDR27
Gene Name
WD repeat domain 27
There are no transcripts associated with this gene.

View Full Product Details