Product Details

SNP ID
rs145215507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3153773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAGTGTGAATTTCTAACAGAGG[C/T]AAAACTGAGCACCATAGTTTACAAG
Phenotype
MIM: 603156 MIM: 615101
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPHL PubMed Links

Gene Details

Gene
BPHL
Gene Name
biphenyl hydrolase-like (serine hydrolase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302777.1 1514 Intron NP_001289706.1
NM_004332.3 1514 Intron NP_004323.2
Gene
TUBB2A
Gene Name
tubulin beta 2A class IIa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001069.2 1514 UTR 3 NP_001060.1
NM_001310315.1 1514 UTR 3 NP_001297244.1

View Full Product Details